Learn More
Enzo Life Sciences Ac-YVAD-pNA (5mg). CAS: 149231-66-3
SDP
Catalog No. 502008233
Encompass_Preferred
Sequence is a modification of pro-interleukin-1β Asp-116 cleavage site for caspase-1. Similar to Ac-YVAD-AMC but cleavage is monitored colorimetrically at 405nm. Alternative name: Caspase-1 substrate (chromogenic). Formula: C29H36N6O10. MW: 628.6. Purity: ≥97% (HPLC). Formulation: Lyophilized. Solubility: Soluble in DMSO. Dilute with buffer pH 7.5. Long Term Storage: -20°C.
Catalog No. 50-200-8233 Supplier Enzo Life Sciences Supplier No. ALX260026M005
Sequence is a modification of pro-interleukin-1β Asp-116 cleavage site for caspase-1. Similar to Ac-YVAD-AMC but cleavage is monitored colorimetrically at 405nm. Alternative name: Caspase-1 substrate (chromogenic). Formula: C29H36N6O10. MW: 628.6. Purity: ≥97% (HPLC). Formulation: Lyophilized. Solubility: Soluble in DMSO. Dilute with buffer pH 7.5. Long Term Storage: -20°C.