Learn More
Sigma Aldrich Fine Chemicals Biosciences AC16 Human Cardiomyocyte Cell Line AC16 Human Cardiomyocytes can be serially passaged and can differentiate when cultured in mitogen-free medium. The cells may be used to study developmental regulation of cardiomyocytes. |
Catalog No. NC2195474
Encompass
AC16 Human Cardiomyocyte Cell Line AC16 Human Cardiomyocytes can be serially passaged and can differentiate when cultured in mitogen-free medium. The cells may be used to study developmental regulation of cardiomyocytes. |
This item is not returnable. View return policy
Catalog No. NC2195474 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SCC109
AC16 Human Cardiomyocyte Cell Line AC16 Human Cardiomyocytes can be serially passaged and can differentiate when cultured in mitogen-free medium. The cells may be used to study developmental regulation of cardiomyocytes. |