Learn More
Medchemexpress LLC HY-100336 1mg Medchemexpress, AC260584 CAS:560083-42-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1003361MG
Encompass_Preferred
Medchemexpress, HY-100336 1mg AC260584 CAS:560083-42-3 AC260584 is an M1 muscarinic receptor allosteric agonist with a pEC 50 of 7.6. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1778