Learn More
TARGETMOL CHEMICALS INC AC4MANNAZ 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1409325MG
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ac4ManNAz is an azido-containing metabolic glycoprotein labeling reagent. It can be used to selectively modify proteins and it can be used in cell labeling tracking and proteomic analysis;purity: 98%
Catalog No. 50-258-1309