Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AirClean™ Systems AC600 Series Vertical Laminar Flow Fume Hood
SDP

Catalog No. 361004380 Shop All AirClean Systems Products
Change view
Click to view available options
Dimensions (D x W x H):
24 x 24 x 30 in.
24 x 32 x 30 in.
24 x 32 x 39 in.
24 x 48 x 32 in.
24 x 48 x 40 in.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (D x W x H)
36-100-4380 24 x 48 x 40 in.
36-100-4377 24 x 32 x 30 in.
36-100-4378 24 x 32 x 39 in.
36-100-4379 24 x 48 x 32 in.
36-100-4376 24 x 24 x 30 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 36-100-4380 Supplier AirClean™ Systems Supplier No. AC648TLFC
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

For applications that are not hazardous to operator but require clean work area, choose an Airclean AC600 Series Vertical Laminar Flow Fume Hood.

 

  • UVTect™ microprocessor controller
  • Class 100 (ISO 5) vertical laminar flow
  • 360-degree visibility
  • Integral polypropylene base for easy cleaning
  • Supplied fully assembled
  • Built-in fluorescent light
  • HEPA filter monitor automatically indicates when filter change is required
  • Available in 110V or 220V versions

 

 

  • Compact, light weight design allows for multiple “clean zones” within laboratory
Options
  • Shortwave UV light
  • Custom sturdy cart allows unit to be made portable; Cart sold separately, see 36-100-0054
  • Polypropylene - metal free construction
  • ULPA filter
  • Sliphatch access port

 

Certifications

UL listed, CSA, CE marked

Compliance

Certified to meet or exceed applicable clean bench standards.

Specifications

Material Thermoplastic
Certifications/Compliance ISO 5 and FS209E, USP 797
Dimensions (D x W x H) 24 x 48 x 40 in.
Voltage 110/220 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.