Learn More
Medchemexpress LLC HY-13493 5mg Medchemexpress, AC710 CAS:1351522-04-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY134935MG
Encompass_Preferred
Medchemexpress, HY-13493 5mg AC710 CAS:1351522-04-7 AC710 is a potent PDGFR inhibitor with Kds of 0.6, 1.57, 1, 1.3, 1.0 nM for FLT3, CSF1R, KIT, PDGFRα and PDGFRβ, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1375