Learn More
Enzo Life Sciences N-(p-Amylcinnamoyl)anthranilic acid (250mg). CAS: 110683-10-8
SDP
Supplier:  Enzo Life Sciences BMLEI1780250
Encompass_Preferred
ACA is a close structural analog of the PLA2 inhibitor, ONO-RS-082 and possesses similar but less potent inhibitory activity. Inhibits TRP channels independently of PLA2, including TRPM2, TRPC6 and TRPM8. It is a useful tool for studying the physiological role of TRP channels. Alternative name: ACA. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO (50 mg/ml) or ethanol (50 mg/ml @ 60°C). Long Term Storage: Ambient.
Catalog No. 50-201-0589