Learn More
Selleck Chemical LLC Acacetin S5318-1g
SDP
Catalog No. 503112770
Encompass_Preferred
Acacetin (Linarigenin 5 7-dihydroxy-4 -methoxyflavone 4 -Methoxyapigenin) is a flavonoid compound that has been shown to have anti-cancer anti-mutagenic anti-inflammatory and anti-peroxidative effects
Catalog No. 50-311-2770 Supplier Selleck Chemical LLC Supplier No. S53181G
Acacetin (Linarigenin 5 7-dihydroxy-4 -methoxyflavone 4 -Methoxyapigenin) is a flavonoid compound that has been shown to have anti-cancer anti-mutagenic anti-inflammatory and anti-peroxidative effects