Learn More
Medchemexpress LLC Acacetin | 480-44-4 | 284.26 | 50 MG
SDP
Catalog No. 5000389096
Encompass_Preferred
Acacetin is an orally active flavonoid, 5,7-Dihydroxy-4'-methoxyflavone, isolated from Dendranthema morifolium. It acts as a PI3Kγ inhibitor, inducing cell cycle arrest, apoptosis, and autophagy in various cancer cells. This compound exhibits potent anti-cancer and anti-inflammatory activities, making it a promising candidate for pain-related disease research.
  • Orally active flavonoid
  • Derived from Dendranthema morifolium
  • Docks in ATP binding pocket of PI3Kγ
  • Causes cell cycle arrest in cancer cells
  • Induces apoptosis and autophagy in cancer cells
  • Potent anti-cancer activity
  • Potent anti-inflammatory activity
  • Potential for pain-related diseases research
  • Suppresses microglial activation in neuroinflammation models
  • Reduces neuronal cell death in ischemia models
  • Decreases visceral and inflammatory nociception
Catalog No. 50-003-89096 Supplier Medchemexpress LLC Supplier No. HYN045150MG
Acacetin is an orally active flavonoid, 5,7-Dihydroxy-4'-methoxyflavone, isolated from Dendranthema morifolium. It acts as a PI3Kγ inhibitor, inducing cell cycle arrest, apoptosis, and autophagy in various cancer cells. This compound exhibits potent anti-cancer and anti-inflammatory activities, making it a promising candidate for pain-related disease research.
  • Orally active flavonoid
  • Derived from Dendranthema morifolium
  • Docks in ATP binding pocket of PI3Kγ
  • Causes cell cycle arrest in cancer cells
  • Induces apoptosis and autophagy in cancer cells
  • Potent anti-cancer activity
  • Potent anti-inflammatory activity
  • Potential for pain-related diseases research
  • Suppresses microglial activation in neuroinflammation models
  • Reduces neuronal cell death in ischemia models
  • Decreases visceral and inflammatory nociception