Learn More
Medchemexpress LLC Acacetin, 5,7-dihydroxy-4'-methoxyflavone | 480-44-4 | 284.26 | 100 MG
SDP
Catalog No. 5000390103
Encompass_Preferred
Acacetin is an orally active flavonoid derived from *Dendranthema morifolium*. It acts by docking in the ATP binding pocket of PI3Kγ. Acacetin induces cell cycle arrest, apoptosis, and autophagy in cancer cells.
  • Possesses potent anti-cancer and anti-inflammatory activity.
  • Has potential for research into pain-related diseases.
  • Decreases cancer cell viabilities in a dose-dependent manner.
  • Induces apoptosis and autophagy.
Catalog No. 50-003-90103 Supplier Medchemexpress LLC Supplier No. HYN0451100MG
Acacetin is an orally active flavonoid derived from *Dendranthema morifolium*. It acts by docking in the ATP binding pocket of PI3Kγ. Acacetin induces cell cycle arrest, apoptosis, and autophagy in cancer cells.
  • Possesses potent anti-cancer and anti-inflammatory activity.
  • Has potential for research into pain-related diseases.
  • Decreases cancer cell viabilities in a dose-dependent manner.
  • Induces apoptosis and autophagy.