Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Acacetin (5,7-dihydroxy-4'-methoxyflavone) | 480-44-4 | MFCD00016936 | 99.7% | 284.26 | C16H12O5 | 10 MG
SDP

Catalog No. 5000296258
Encompass_Preferred

Acacetin (5,7-dihydroxy-4'-methoxyflavone) is an O-methylated flavone used as a research reagent for studies of apoptosis, autophagy, and inflammatory signaling. It is supplied as a high-purity reference material for in vitro and preclinical experiments.

  • CAS number: 480-44-4.
  • Molecular formula: C16H12O5.
  • Molecular weight: 284.26 g/mol.
  • Purity: 99.7%.
  • Available pack sizes: 1 mg-1 g for laboratory use.
  • Documentation provided: data sheet, certificate of analysis, and safety data sheet.

Catalog No. 50-002-96258 Supplier Medchemexpress LLC Supplier No. HYN045110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acacetin (5,7-dihydroxy-4'-methoxyflavone) is an O-methylated flavone used as a research reagent for studies of apoptosis, autophagy, and inflammatory signaling. It is supplied as a high-purity reference material for in vitro and preclinical experiments.

  • CAS number: 480-44-4.
  • Molecular formula: C16H12O5.
  • Molecular weight: 284.26 g/mol.
  • Purity: 99.7%.
  • Available pack sizes: 1 mg-1 g for laboratory use.
  • Documentation provided: data sheet, certificate of analysis, and safety data sheet.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.