Learn More
TARGETMOL CHEMICALS INC Acacetin 50MG
SDP
Catalog No. 502879174
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Acacetin (5,7-Dihydroxy-4'-methoxyflavone) is an O-methylated flavone found in various plants, exhibiting antinociceptive, anti-inflammatory, and antioxidant activities in different research models. Purity 98%
Catalog No. 50-287-9174 Supplier TARGETMOL CHEMICALS INC Supplier No. T398150MG
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Acacetin (5,7-Dihydroxy-4'-methoxyflavone) is an O-methylated flavone found in various plants, exhibiting antinociceptive, anti-inflammatory, and antioxidant activities in different research models. Purity 98%