Learn More
Medchemexpress LLC Acacetin-7-O-β-D-galactopyranoside | 80443-15-8 | 98.6% | 446.40 | C22H22O10 | 5mg
SDP
Catalog No. 5000269619
Encompass_Preferred
Acacetin-7-O- -D-galactopyranoside is a flavonoid that can be isolated from flower heads of Chrysanthemum morifolium Acacetin-7-O- -D-galactopyranoside inhibits AChE activity and can be used for research of Alzheimer s disease[1]
Catalog No. 50-002-69619 Supplier Medchemexpress LLC Supplier No. HYW7425645MG
Acacetin-7-O- -D-galactopyranoside is a flavonoid that can be isolated from flower heads of Chrysanthemum morifolium Acacetin-7-O- -D-galactopyranoside inhibits AChE activity and can be used for research of Alzheimer s disease[1]