Learn More
Aviva Systems Biology Corp ACADM Antibody - N-terminal region (ARP32788_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32788_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ACADM. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ACADM. Alternate names: ACAD1, MCAD, MCADHApplicationWB. Species reactivity: Cow, Horse, Human, Mouse, Rat
Catalog No. 50-014-54765