Learn More
Aviva Systems Biology Corp ACADSB Antibody - middle region (ARP54586_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54586_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ACADSB. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ACADSB. Alternate names: 2-MEBCAD, ACAD7, SBCADApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-61250