Learn More
Cayman Chemical AcalabrutInIb 10mg
SDP
Catalog No. 502981150
Encompass_Preferred
An irreversible BTK inhibitor; dose-dependently inhibits B cell receptor signaling in primary CLL cells
Catalog No. 50-298-1150 Supplier Cayman Chemical Supplier No. 1989910 MG
An irreversible BTK inhibitor; dose-dependently inhibits B cell receptor signaling in primary CLL cells