Learn More
Selleck Chemical LLC Acalabrutinib
SDP
Catalog No. 5000000825
Encompass_Preferred
Acalabrutinib (ACP-196) is a selective second-generation Bruton s tyrosine kinase (BTK) inhibitor with an IC50 of 3 nM which prevents the activation of the B-cell antigen receptor (BCR) signaling pathway ACP-196 has improved target specificity with 323- 94- 19- and 9-fold selectivity over the other TEC kinase family members (ITK TXK BMX and TEC respectively) and no activity against EGFR
Catalog No. 50-000-00825 Supplier Selleck Chemical LLC Supplier No. S8116100MG
Acalabrutinib (ACP-196) is a selective second-generation Bruton s tyrosine kinase (BTK) inhibitor with an IC50 of 3 nM which prevents the activation of the B-cell antigen receptor (BCR) signaling pathway ACP-196 has improved target specificity with 323- 94- 19- and 9-fold selectivity over the other TEC kinase family members (ITK TXK BMX and TEC respectively) and no activity against EGFR