Learn More
Selleck Chemical LLC Acalabrutinib (ACP-196) 5mg 1420477-60-6
SDP
Supplier:  Selleck Chemical LLC S81165MG
Encompass_Preferred
Acalabrutinib (ACP-196) is a selective second-generation Bruton's tyrosine kinase (BTK) inhibitor with an IC50 of 3 nM, which prevents the activation of the B-cell antigen receptor (BCR) signaling pathway. ACP-196 has improved target specificity over ibrutinib with 323-, 94-, 19- and 9-fold selectivity over the other TEC kinase family members (ITK, TXK, BMX, and TEC, respectively) and no activity against EGFR. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1142630