Learn More
Selleck Chemical LLC acalisib
SDP
Catalog No. 5000003743
Encompass_Preferred
Acalisib (GS-9820 CAL-120) is a highly selective and potent p110 inhibitor (IC50 14 nM) with 114- to 400-fold selectivity over the other class I PI3K enzymes and no activity against Class II and III PI3K family members or other PI3K-related proteins including mTOR and DNA-PK
Catalog No. 50-000-03743 Supplier Selleck Chemical LLC Supplier No. S58185MG
Acalisib (GS-9820 CAL-120) is a highly selective and potent p110 inhibitor (IC50 14 nM) with 114- to 400-fold selectivity over the other class I PI3K enzymes and no activity against Class II and III PI3K family members or other PI3K-related proteins including mTOR and DNA-PK