Learn More
Selleck Chemical LLC Acamprosate Calcium S3748-5mg
SDP
Catalog No. 503108972
Encompass_Preferred
Acamprosate (N-Acetylhomotaurine Calcium acetylhomotaurinate N-acetylhomotaurinate) is a synthetic compound with a chemical structure similar to the amino acid neurotransmitter gamma-aminobutyric acid (GABA) and the amino acid neuromodulator taurine
Catalog No. 50-310-8972 Supplier Selleck Chemical LLC Supplier No. S37485MG
Acamprosate (N-Acetylhomotaurine Calcium acetylhomotaurinate N-acetylhomotaurinate) is a synthetic compound with a chemical structure similar to the amino acid neurotransmitter gamma-aminobutyric acid (GABA) and the amino acid neuromodulator taurine