Learn More
Medchemexpress LLC Acamprosate-d3 calcium | 1225580-94-8 | 98.0% | 203.26 | 1 MG
SDP
Catalog No. 5000385705
Encompass_Preferred
Acamprosate-d3 (calcium) is the deuterium labeled Acamprosate calcium. Acamprosate calcium is a GABA receptor agonist and modulator of glutamatergic systems. It is for research use only and not sold to patients.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-85705 Supplier Medchemexpress LLC Supplier No. HY17030S1MG
Acamprosate-d3 (calcium) is the deuterium labeled Acamprosate calcium. Acamprosate calcium is a GABA receptor agonist and modulator of glutamatergic systems. It is for research use only and not sold to patients.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS