Learn More
Medchemexpress LLC Acamprosate-d3 calcium | 1225580-94-8 | 98.0% | 203.26 | 5 MG
SDP
Catalog No. 5000385796
Encompass_Preferred
Acamprosate-d3 (calcium) is the deuterium-labeled form of Acamprosate calcium. Acamprosate calcium functions as a GABA receptor agonist and modulates glutamatergic systems. This compound is intended for research use only and is not sold to patients.
  • Deuterium labeled Acamprosate calcium
  • GABA receptor agonist
  • Modulator of glutamatergic systems
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes are incorporated into drug molecules for quantitation during drug development
  • Deuteration can impact pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-85796 Supplier Medchemexpress LLC Supplier No. HY17030S5MG
Acamprosate-d3 (calcium) is the deuterium-labeled form of Acamprosate calcium. Acamprosate calcium functions as a GABA receptor agonist and modulates glutamatergic systems. This compound is intended for research use only and is not sold to patients.
  • Deuterium labeled Acamprosate calcium
  • GABA receptor agonist
  • Modulator of glutamatergic systems
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes are incorporated into drug molecules for quantitation during drug development
  • Deuteration can impact pharmacokinetic and metabolic profiles of drugs