Learn More
Cayman Chemical AcamprosatecalcIum salt 100mg
SDP
Catalog No. 502994341
Encompass_Preferred
An NMDA modulator with antagonist or agonist effects depending on brain region, receptor composition, and other factors; an acetylated derivative of homotaurine; decreases paired-pulse inhibition of GABAA IPSCs, indicating that it may inhibit GABAB autoreceptor-mediated inhibition of GABA release (300 µM); reduces voluntary intake of ethanol in rats (0.26 and 0.52 mmol/kg per day, i.p.)
Catalog No. 50-299-4341 Supplier Cayman Chemical Supplier No. 23899100 MG
An NMDA modulator with antagonist or agonist effects depending on brain region, receptor composition, and other factors; an acetylated derivative of homotaurine; decreases paired-pulse inhibition of GABAA IPSCs, indicating that it may inhibit GABAB autoreceptor-mediated inhibition of GABA release (300 µM); reduces voluntary intake of ethanol in rats (0.26 and 0.52 mmol/kg per day, i.p.)