Learn More
Cayman Chemical Acamprosate (calcium salt), Purity: ≥98%, CAS: 77337-73-6, 500 mg
SDP
Catalog No. 502995314
Encompass_Preferred
An NMDA modulator with antagonist or agonist effects depending on brain region, receptor composition, and other factors; an acetylated derivative of homotaurine; decreases paired-pulse inhibition of GABAA IPSCs, indicating that it may inhibit GABAB autoreceptor-mediated inhibition of GABA release (300 µM); reduces voluntary intake of ethanol in rats (0.26 and 0.52 mmol/kg per day, i.p.)
Catalog No. 50-299-5314 Supplier Cayman Chemical Supplier No. 23899500 MG
An NMDA modulator with antagonist or agonist effects depending on brain region, receptor composition, and other factors; an acetylated derivative of homotaurine; decreases paired-pulse inhibition of GABAA IPSCs, indicating that it may inhibit GABAB autoreceptor-mediated inhibition of GABA release (300 µM); reduces voluntary intake of ethanol in rats (0.26 and 0.52 mmol/kg per day, i.p.)