Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Aggrecan (ACAN) protein | 20UG
SDP

Catalog No. 5000177112
Encompass_Preferred

Recombinant human aggrecan (ACAN) protein comprising amino acids A17-L721, expressed in HEK293 cells and supplied as a lyophilized, His-tagged preparation for research use. The material is formulated in PBS (pH 7.4) with 8% trehalose, has an estimated molecular weight of 90-130 kDa, and endotoxin <1 EU/μg. Store lyophilized at -20°C; reconstitute to ≥100 μg/mL as recommended.

  • Recombinant human ACAN spanning A17-L721.
  • Expressed in HEK293 cells with C-terminal His tag.
  • Lyophilized from PBS (pH 7.4) with 8% trehalose.
  • Molecular weight approximately 90-130 kDa.
  • Endotoxin level less than 1 EU/μg (LAL method).
  • Store at -20°C; freeze aliquots at -20°C or -80°C for extended storage.
  • Intended for research use only.

Catalog No. 50-001-77112 Supplier Medchemexpress LLC Supplier No. HYP70105620UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human aggrecan (ACAN) protein comprising amino acids A17-L721, expressed in HEK293 cells and supplied as a lyophilized, His-tagged preparation for research use. The material is formulated in PBS (pH 7.4) with 8% trehalose, has an estimated molecular weight of 90-130 kDa, and endotoxin <1 EU/μg. Store lyophilized at -20°C; reconstitute to ≥100 μg/mL as recommended.

  • Recombinant human ACAN spanning A17-L721.
  • Expressed in HEK293 cells with C-terminal His tag.
  • Lyophilized from PBS (pH 7.4) with 8% trehalose.
  • Molecular weight approximately 90-130 kDa.
  • Endotoxin level less than 1 EU/μg (LAL method).
  • Store at -20°C; freeze aliquots at -20°C or -80°C for extended storage.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.