Learn More
Medchemexpress LLC Acanthoside B | 7374-79-0 | 5 MG
SDP
Catalog No. 5000420771
Encompass_Preferred
Acanthoside B is a potential bioactive lignan with anti-inflammatory and anti-amnesic activities, making it suitable for research into Alzheimer's disease and lung inflammation. It has been shown to attenuate scopolamine-inflicted AD-like amnesic traits in mice.
  • Potential bioactive lignan
  • Exhibits anti-inflammatory activities
  • Exhibits anti-amnesic activities
  • Applicable for Alzheimer's disease research
  • Applicable for lung inflammation research
  • Attenuates scopolamine-inflicted AD-like amnesic traits
  • Restores cholinergic activity
  • Decreases endogenous antioxidant status
  • Suppresses neuroinflammation
  • Activates the TrkB/CREB/BDNF pathway
Catalog No. 50-004-20771 Supplier Medchemexpress LLC Supplier No. HYN28075MG
Acanthoside B is a potential bioactive lignan with anti-inflammatory and anti-amnesic activities, making it suitable for research into Alzheimer's disease and lung inflammation. It has been shown to attenuate scopolamine-inflicted AD-like amnesic traits in mice.
  • Potential bioactive lignan
  • Exhibits anti-inflammatory activities
  • Exhibits anti-amnesic activities
  • Applicable for Alzheimer's disease research
  • Applicable for lung inflammation research
  • Attenuates scopolamine-inflicted AD-like amnesic traits
  • Restores cholinergic activity
  • Decreases endogenous antioxidant status
  • Suppresses neuroinflammation
  • Activates the TrkB/CREB/BDNF pathway