Learn More
Medchemexpress LLC Acanthoside B | 7374-79-0 | 99.5% | 580.58 g/mol | C28H36O13 | 10 MG
SDP
Catalog No. 5000289075
Encompass_Preferred
Acanthoside B is a bioactive lignan supplied for laboratory research with reported anti-inflammatory and anti-amnesic activities. It is used in preclinical studies investigating Alzheimer's disease mechanisms and lung inflammation models.
  • High purity suitable for research (approximately 99.5%).
  • Useful in anti-inflammatory and anti-amnesic studies.
  • Applicable to Alzheimer's disease and lung inflammation research.
  • Molecular formula C28H36O13, molecular weight 580.58 g/mol.
  • Supplied as 10 mg packages with recommended storage at -20°C (in solvent: -80°C for long-term storage).
Catalog No. 50-002-89075 Supplier Medchemexpress LLC Supplier No. HYN280710MG
Acanthoside B is a bioactive lignan supplied for laboratory research with reported anti-inflammatory and anti-amnesic activities. It is used in preclinical studies investigating Alzheimer's disease mechanisms and lung inflammation models.
  • High purity suitable for research (approximately 99.5%).
  • Useful in anti-inflammatory and anti-amnesic studies.
  • Applicable to Alzheimer's disease and lung inflammation research.
  • Molecular formula C28H36O13, molecular weight 580.58 g/mol.
  • Supplied as 10 mg packages with recommended storage at -20°C (in solvent: -80°C for long-term storage).