Learn More
Apexbio Technology LLC Acarbose 56180-94-0 1g
SDP
Catalog No. 5000062939
Encompass_Preferred
Acarbose (CAS 56180-94-0) is a small-molecule inhibitor targeting intestinal -glucosidase an enzyme involved in carbohydrate digestion It exerts its pharmacological action by hindering the hydrolytic breakdown of complex carbohydrates into monosaccharides thereby slowing glucose absorption Demonstrating potent inhibitory activity with an IC of approximately 11 nM acarbose is widely explored in biomedical research contexts related to glucose regulation and diabetes management
Catalog No. 50-000-62939 Supplier Apexbio Technology LLC Supplier No. A24311000
Acarbose (CAS 56180-94-0) is a small-molecule inhibitor targeting intestinal -glucosidase an enzyme involved in carbohydrate digestion It exerts its pharmacological action by hindering the hydrolytic breakdown of complex carbohydrates into monosaccharides thereby slowing glucose absorption Demonstrating potent inhibitory activity with an IC of approximately 11 nM acarbose is widely explored in biomedical research contexts related to glucose regulation and diabetes management