Learn More
Medchemexpress LLC HY-B0089 200mg Medchemexpress, Acarbose CAS:56180-94-0 Purity:>98%
SDP
Catalog No. 502082358
Encompass_Preferred
Medchemexpress, HY-B0089 200mg Acarbose CAS:56180-94-0 Acarbose (BAY g 5421), antihyperglycemic agent, is an orally active alpha-glucosidase inhibitor (IC50=11 nM). Acarbose can potentiate the hypoglycemic effects of sulfonylureas or insulin. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2358 Supplier Medchemexpress LLC Supplier No. HYB0089200MG
Medchemexpress, HY-B0089 200mg Acarbose CAS:56180-94-0 Acarbose (BAY g 5421), antihyperglycemic agent, is an orally active alpha-glucosidase inhibitor (IC50=11 nM). Acarbose can potentiate the hypoglycemic effects of sulfonylureas or insulin. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.