Learn More
Medchemexpress LLC Acarbose | 56180-94-0 | 99.97% | C25H43NO18 | 10 MM/1 ML
SDP
Catalog No. 5000432525
Encompass_Preferred
Acarbose is an orally active alpha-glucosidase inhibitor and an antihyperglycemic agent. It works by slowing the digestion and absorption of complex carbohydrates, thereby leading to a more gradual rise in blood glucose levels after meals. Primarily used in the management of type 2 diabetes mellitus, it can potentiate the hypoglycemic effects of sulfonylureas or insulin.
  • Antihyperglycemic agent
  • Orally active alpha-glucosidase inhibitor (IC50=11 nM)
  • Potentiates hypoglycemic effects of sulfonylureas or insulin
  • Slows digestion and absorption of carbohydrates
  • Used for management of type 2 diabetes mellitus
  • Available as a solid, white to light yellow powder
  • Soluble in water
Catalog No. 50-004-32525 Supplier Medchemexpress LLC Supplier No. HYB008910MM/1ML
Acarbose is an orally active alpha-glucosidase inhibitor and an antihyperglycemic agent. It works by slowing the digestion and absorption of complex carbohydrates, thereby leading to a more gradual rise in blood glucose levels after meals. Primarily used in the management of type 2 diabetes mellitus, it can potentiate the hypoglycemic effects of sulfonylureas or insulin.
  • Antihyperglycemic agent
  • Orally active alpha-glucosidase inhibitor (IC50=11 nM)
  • Potentiates hypoglycemic effects of sulfonylureas or insulin
  • Slows digestion and absorption of carbohydrates
  • Used for management of type 2 diabetes mellitus
  • Available as a solid, white to light yellow powder
  • Soluble in water