Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acarbose Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014624 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4624 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4624 Supplier MilliporeSigma Supelco Supplier No. PHR1253500MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Acarbose is an oral antidiabetic drug, a glucosidase inhibitor used in the management of non-insulin-dependent diabetes mellitus (NIDDM). Featured Industries: Food and Beverages Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 56180-94-0
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C25H43 NO18
MDL Number MFCD00869592
Synonym Acarbose; 4 ′′,6 ′′-Dideoxy-4 ′′-([1 S]-[1,4,6/5 ]-4,5,6-trihydroxy-3-hydroxymethyl-2-yclohexenylamino)-maltotriose
Type Certified Reference Material
Quantity 500 mg
Formula Weight 645.60
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.