Learn More
Cayman Chemical aCardIac ActInTerm Rabb 100uL
SDP
Catalog No. 502947535
Encompass_Preferred
An isomer of vitamin D3 increases tibia ash weight and bone mineralization in vitamin D-deficient chicks at 1 3 and 10 Ug/day induces intestinal calcium transport and bone calcium mobilization in anephric rats fed a low-calcium and vitamin D-deficient diet at 25 Ug/animal
Catalog No. 50-294-7535 Supplier Cayman Chemical Supplier No. 32211100 UL
An isomer of vitamin D3 increases tibia ash weight and bone mineralization in vitamin D-deficient chicks at 1 3 and 10 Ug/day induces intestinal calcium transport and bone calcium mobilization in anephric rats fed a low-calcium and vitamin D-deficient diet at 25 Ug/animal