Learn More
Aviva Systems Biology Corp ACAT2 Antibody - middle region (ARP32790_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32790_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ACAT2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human ACAT2. Alternate names: ACAT2, ACTLApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54767