Learn More
Aviva Systems Biology Corp Acbd3 Antibody - middle region (ARP57650_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57650_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Acbd3. It was validated on Western Blot. . Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-286-341