Learn More
Aviva Systems Biology Corp ACBD3 Antibody - N-terminal region (ARP57649_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57649_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ACBD3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ACBD3. Alternate names: GCP60, GOCAP1, GOLPH1, PAP7ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62914