Learn More
Aviva Systems Biology Corp ACBD6 Antibody - middle region (ARP60392_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60392_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ACBD6. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human ACBD6. Alternate names: MGC2404ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit
Catalog No. 50-287-029