Learn More
Aviva Systems Biology Corp ACBD7 Antibody - N-terminal region (ARP56265_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56265_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ACBD7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ACBD7. Alternate names: FLJ38219, MGC33893, bA455B2.2ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61788