Learn More
Medchemexpress LLC HY-128359 5mg , ACBI1 CAS:2375564-55-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1283595MG
Encompass_Preferred
HY-128359 5mg ACBI1 CAS: 2375564-55-7 ACBI1 is a potent and cooperative SMARCA2, SMARCA4 and PBRM1 degrader with DC50s of 6, 11 and 32 nM, respectively. ACBI1 is a PROTAC degrader. ACBI1 shows anti-proliferative activity. ACBI1 induces apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-8980