Learn More
New England Biolabs, Inc. Acc65I – 2000 units
SDP
Catalog No. 50812320
Encompass
A restriction endonuclease that recognizes the sequence G^GTAC_C.
Catalog No. 50-812-320 Supplier New England Biolabs, Inc. Supplier No. R0599S
A restriction endonuclease that recognizes the sequence G^GTAC_C.