Learn More
Andwin Scientific BLADE ACCU-EDGE MICROTOME 50/pk
SDP
Catalog No. 50465338
Encompass_Preferred
The Accu-Edge disposable low-profile blades by Sakura Finetek are compatible with all Accu-Edge blade holders. This product is ideal for microtomy, cryotomy, and sectioning specimens without striations, distortion, or chattering. It is sold in dispensers that can store 50 blades and distribute them in a few seconds. The previously used blade is then automatically ejected.
This item is not returnable. View return policy
Catalog No. 50-465-338 Supplier Andwin Scientific Supplier No. 4689
The Accu-Edge disposable low-profile blades by Sakura Finetek are compatible with all Accu-Edge blade holders. This product is ideal for microtomy, cryotomy, and sectioning specimens without striations, distortion, or chattering. It is sold in dispensers that can store 50 blades and distribute them in a few seconds. The previously used blade is then automatically ejected.