Learn More
GSC Go Science Crazy Acceleration Trolley
Catalog No. S28107 Shop All GSC Products
Change view
Click to view available options
50mm pulleys; attachment kit with hardware
Catalog No.
S28107 50mm pulleys; attachment kit with hardware
1 options
Catalog No. S28107 Supplier GSC Go Science Crazy Supplier No. ACT01/DEL
Demonstrate that the rate of acceleration of an object depends on the angle of incline. The GSC International Acceleration Trolley has 3 holes for attaching weights and brackets to place on the cable at any point. Includes a trolley with 50mm diameter pulleys, wire cable, attachment kit with hardware and instructions.
  • Demonstrate that the rate of acceleration of an object depends on the angle of incline
  • Trolley has 3 holes for attaching weights and brackets to place on the cable at any point
  • A mass set is also needed for the demonstration
Specifications
Includes Includes trolley with 50mm diameter pulleys, wire cable, attachment kit with hardware and instructions