Learn More
HORIZON DISCOVERY BIOSCIENCES LIMITED Molecular Grade RNase-free water. 100ml
Catalog No. NC0693367
Encompass
Molecular Grade water for dilution of 5x siRNA Buffer or resuspension of RNA. RNase-free to prevent degradation of RNA reagents and oligonucleotides.
This item is not returnable. View return policy
Catalog No. NC0693367 Supplier HORIZON DISCOVERY BIOSCIENCES LIMITED Supplier No. B003000WB100
Molecular Grade water for dilution of 5x siRNA Buffer or resuspension of RNA. RNase-free to prevent degradation of RNA reagents and oligonucleotides.