Learn More
HORIZON DISCOVERY BIOSCIENCES LIMITED Accell Human SLC3A2 (6520) siRNA - SMARTpool 5 nmol
Catalog No. NC3452585
Encompass
Accell siRNA offers what no other RNAi product can claim: delivery into difficult-to-transfect cells without transfection reagents, virus, or instrumentation. Achieve targeted gene silencing in neuronal, immune, or primary cells that are beyond the reach of conventional RNAi products due to toxicity caused by transfection reagents or undesirable viral responses.
Catalog No. NC3452585 Supplier HORIZON DISCOVERY BIOSCIENCES LIMITED Supplier No. E003542000005
Accell siRNA offers what no other RNAi product can claim: delivery into difficult-to-transfect cells without transfection reagents, virus, or instrumentation. Achieve targeted gene silencing in neuronal, immune, or primary cells that are beyond the reach of conventional RNAi products due to toxicity caused by transfection reagents or undesirable viral responses.