Learn More
HORIZON DISCOVERY BIOSCIENCES LIMITED Accell Human SLC7A5 (8140) siRNA - SMARTpool 5 nmol
Catalog No. NC3452571
Encompass
A novel siRNA for difficult-to-transfect cells, modified to require no transfection reagent or viral vector for delivery. Predesigned Accell siRNA is available as individual reagents and in SMARTpool format.
Catalog No. NC3452571 Supplier HORIZON DISCOVERY BIOSCIENCES LIMITED Supplier No. E004953000005
A novel siRNA for difficult-to-transfect cells, modified to require no transfection reagent or viral vector for delivery. Predesigned Accell siRNA is available as individual reagents and in SMARTpool format.