Learn More
HORIZON DISCOVERY BIOSCIENCES LIMITED Accell siRNA Reagents - Human, SMARTPool, 20 nmol
Catalog No. NC1859613
Encompass
A mixture of 4 siRNA provided as a single reagent, providing advantages in both potency and specificity.
Catalog No. NC1859613 Supplier HORIZON DISCOVERY BIOSCIENCES LIMITED Supplier No. E003808000020
A mixture of 4 siRNA provided as a single reagent, providing advantages in both potency and specificity.