Learn More
HORIZON DISCOVERY BIOSCIENCES LIMITED Horizon Discovery Biosciences Accell siRNA, Human, WDFY3, 50 nmol
Catalog No. NC3919085
Encompass
This product consists of Accell siRNA targeting the human WDFY3 gene. Accell siRNA is designed for efficient delivery into difficult-to-transfect cell types without the requirement for transfection reagents or viral vectors.
  • Provides reliable gene knockdown in challenging primary and suspension cells
  • Features proprietary modifications to improve uptake and stability
  • Optimized for direct addition to cell culture media
  • Validated for effective reduction of target gene expression
Catalog No. NC3919085 Supplier HORIZON DISCOVERY BIOSCIENCES LIMITED Supplier No. E012924000050
This product consists of Accell siRNA targeting the human WDFY3 gene. Accell siRNA is designed for efficient delivery into difficult-to-transfect cell types without the requirement for transfection reagents or viral vectors.
  • Provides reliable gene knockdown in challenging primary and suspension cells
  • Features proprietary modifications to improve uptake and stability
  • Optimized for direct addition to cell culture media
  • Validated for effective reduction of target gene expression