Learn More
HORIZON DISCOVERY BIOSCIENCES LIMITED Accell Non-targeting Control siRNA #1, 20nmol
Catalog No. NC1567415
Encompass
A negative control siRNA to determine baseline cellular response to Accell siRNA application. Accell siRNA requires no transfection reagent or viral vector for delivery into difficult-to-transfect cells.
Catalog No. NC1567415 Supplier HORIZON DISCOVERY BIOSCIENCES LIMITED Supplier No. D0019100120
A negative control siRNA to determine baseline cellular response to Accell siRNA application. Accell siRNA requires no transfection reagent or viral vector for delivery into difficult-to-transfect cells.