Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp AccellPlus CM Ion Exchange Packing, 40 μm, 300 Å, Polymer-Coated Silica, Weak Cation Exchanger

Catalog No. 50784983 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

Accell Plus ion exchange packings are polymer-coated, silica-based materials designed for preparative isolations based on ionic interactions, particularly for proteins, enzymes, and immunoglobulins. This specific product, AccellPlus CM, is a weak cation exchanger.

  • Particle size: 40 μm
  • Pore size: 300 Å
  • Material type: Polymer-coated silica
  • Chemistry: Cation-exchange
  • Separation mode: Ion-exchange
  • Molecular weight range: 10,000 to 500,000
  • pH range: 2 to 9
  • Silanol activity: Low
  • Particle shape: Spherical
  • Mass: 100 g
  • Carbon load: 5.5%
  • Application: Protein

Catalog No. 50-784-983 Supplier Waters Corp Supplier No. WAT010740
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Accell Plus ion exchange packings are polymer-coated, silica-based materials designed for preparative isolations based on ionic interactions, particularly for proteins, enzymes, and immunoglobulins. This specific product, AccellPlus CM, is a weak cation exchanger.

  • Particle size: 40 μm
  • Pore size: 300 Å
  • Material type: Polymer-coated silica
  • Chemistry: Cation-exchange
  • Separation mode: Ion-exchange
  • Molecular weight range: 10,000 to 500,000
  • pH range: 2 to 9
  • Silanol activity: Low
  • Particle shape: Spherical
  • Mass: 100 g
  • Carbon load: 5.5%
  • Application: Protein

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.