Learn More
Waters Corp Waters AccellPlus Ion Exchange Packing, 40 μm, 300 Å, Cation-Exchange, 500 g
Catalog No. 50784982 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
AccellPlus ion exchange packings are 40μm, 300 Å polymer-coated silica-based materials designed for preparative isolations based on ionic interactions. These packings are particularly suited for proteins, enzymes, and immunoglobulins, and can be used in Advanced Purification (AP) glass columns.
  • 40μm, 300 Å polymer-coated silica-based material
  • Weak cation exchanger chemistry
  • Suitable for preparative isolations based on ionic interactions
  • Optimized for proteins, enzymes, and immunoglobulins
  • Can be packed into Advanced Purification (AP) glass columns
  • Low silanol activity
  • Spherical particle shape
  • pH range of 2 to 9 pH
Catalog No. 50-784-982 Supplier Waters Corp Supplier No. WAT010739
AccellPlus ion exchange packings are 40μm, 300 Å polymer-coated silica-based materials designed for preparative isolations based on ionic interactions. These packings are particularly suited for proteins, enzymes, and immunoglobulins, and can be used in Advanced Purification (AP) glass columns.
  • 40μm, 300 Å polymer-coated silica-based material
  • Weak cation exchanger chemistry
  • Suitable for preparative isolations based on ionic interactions
  • Optimized for proteins, enzymes, and immunoglobulins
  • Can be packed into Advanced Purification (AP) glass columns
  • Low silanol activity
  • Spherical particle shape
  • pH range of 2 to 9 pH