Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Corporation Bulk Media, 100 g, 40 μm, Anion-Exchange

Catalog No. 50784985 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

These ion exchange packings are 40μm, 300 angstrom polymer-coated silica-based materials, designed for preparative isolations of proteins, enzymes, and immunoglobulins. Available in strong anion and weak cation exchangers, they can be packed into advanced purification glass columns.

  • Mass of 100 g.
  • Anion-exchange chemistry.
  • Ion-exchange separation mode.
  • Molecular weight range of 10,000 to 500,000.
  • Spherical particle shape.
  • Low silanol activity.
  • pH range of 2 to 9.
  • Application for protein.
  • Supplied as 1 unit per package.

Catalog No. 50-784-985 Supplier Waters Corp Supplier No. WAT010742
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These ion exchange packings are 40μm, 300 angstrom polymer-coated silica-based materials, designed for preparative isolations of proteins, enzymes, and immunoglobulins. Available in strong anion and weak cation exchangers, they can be packed into advanced purification glass columns.

  • Mass of 100 g.
  • Anion-exchange chemistry.
  • Ion-exchange separation mode.
  • Molecular weight range of 10,000 to 500,000.
  • Spherical particle shape.
  • Low silanol activity.
  • pH range of 2 to 9.
  • Application for protein.
  • Supplied as 1 unit per package.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.